Learn More
Microsolv Technology Corporation Union, Connector, Tee, High Pressure, 0.50mm Through Holes with 10-32 Screw Threads. Carbon PEEK. Use with 1/16" Tubing. 1 EA.
Supplier:  Microsolv Technology Corporation 4730211
Encompass_Preferred
This "Union" is used to connect the same size PEEK or Stainless Steel tubing on all ends, in the high pressures zones of HPLC systems. A "Tee Connection" is often used to split flow into two directions or to combine two streams into one. Carbon PEEK has been reinforced with carbon fiber, giving it the highest strength and stiffness of all the PEEK grades and improving its load-carrying capacity.
Catalog No. 50-227-1451