Learn More
IDEX Health & Science Upchurch UNION PEEK LRG BORE 3/16IN TUB
Supplier:  IDEX Health & Science Upchurch P134
Encompass_Preferred
Idex Peek Low-Pressure Union Assembly, Natural, For Use With: 5/16-24, Length: 0.78 in., PEEK, Width: 0.44 in.
Catalog No. NC1920538