Learn More
Med Vet International Uniprim Powder Packet, 67/333mg, 37.5gm
Supplier:  Med Vet International RXUNIPRIM
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Uniprim powder for horses contains 67mg trimethoprim and 333mg sulfadiazine per gram that helps to provide effective antibacterial activity against a wide range of bacterial infections in animals.
Catalog No. 50-282-8410