Learn More
Med Vet International Uniprim Powder For Horses, 400gm
Supplier:  Med Vet International RXUNIPRIM400
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Trimethoprim/sulfadiazine is indicated in horses where potent systemic antibacterial action against sensitive organisms is required. Trimethoprim/sulfadiazine is indicated where control of bacterial infections is required during treatment of: Acute Strangles, Respiratory Tract Infections, Acute Urogenital Infections, Wound Infections and Abscesses. Trimethoprim/sulfadiazine is well tolerated by foals. See package insert for complete product details.
Catalog No. 50-282-8411