Learn More
Electron Microscopy Sciences Corning Digital Stirring Hot Plate Universal Accessory Kit
SDP
Supplier:  Electron Microscopy Sciences 7235150
Encompass_Preferred
Kit is for all Corning digital display hot plates and stirring hot plates (Models PC-400D, PC-420D, PC600D, PC-620D). It is voltage independent and includes a Temperature Controller (6795PR), a two piece, 5/16" diameter, 18".
Catalog No. 50-189-9847