Learn More
Perkin Elmer US LLC Universal ATR Shoe - 0.5 mm
Supplier:  Perkin Elmer US LLC L1202049
Encompass_Preferred
PerkinElmer's Universal ATRs are excellent, easy-to-use accessories for the analysis of solids and liquids. These instruments produce high quality spectra through the use of a pressure arm, ensuring sample to sample and operator to operator reproducibility.
Catalog No. 50-211-1755