Learn More
Biochain Institute Inc Universal cDNA Reverse Transcribed by Oligo dT Primer: Dog Normal Tissues, 100 rxn/PK
SDP
Supplier:  Biochain Institute Inc C4734565
Encompass_Preferred
BioChain's Dog Universal cDNA is prepared from major organs of both male and female Beagle dogs. This cDNA serves as a standard for comparison of gene expression by RT-PCR or as a gene pool for cloning dog genes.
Catalog No. 50-184-211