Learn More
Biochain Institute Inc Universal cDNA Reverse Transcribed by Random Hexamer: Rat Normal Tissues, 100 rxn/PK
SDP
Supplier:  Biochain Institute Inc C4434567R
Encompass_Preferred
BioChains' Rat Universal cDNA is prepared from several male and female Sprague Dawley or Wistar rat whole bodies without fur. This cDNA serves as a standard for comparison of gene expression by RT-PCR or as a gene pool for cloning rat genes.
Catalog No. 50-184-220