Learn More
Med Vet International Universal "F" Flex Circuit 38"-108"w/o bag, J0248WA, Each
Supplier:  Med Vet International JOR248WA
Encompass_Preferred
The corrugated tubing of this unilimb circuit offers expandable and contraction capability. The clinician can determine the length of the hose needed for every surgery. Better positioning - the hose can be set at varying angles versus just standard straight on tubing. Excess tubing can be contracted and out of the way. Includes right angle elbow with gas port. Flex Circuit. 108" expanded length to 38" contracted length.
Catalog No. 50-005-81582