Learn More
MSC Proto Universal Joint, 1/2 Drive, 2.75 in L, 1 ea
Supplier:  MSC 38234100
Encompass
This universal joint is designed for mechanical applications, featuring 1/2 in male and 1/2 in female drive sizes for versatility in various tasks.
  • Ball and spring socket retention
  • Chrome plating provides increased protection from rust and corrosion
  • Spinner disk is ideal for fast turn down of fasteners
  • Meets ANSI Spec B107.1
  • Meets Federal Spec GGG-W-641e
  • Manufactured in the United States
Catalog No. 50-192-0194