Learn More
Mascon Medical Mascon Medical™ Universal Pipette Tips, 100μl, 96 Tips/Rack, 10 Racks/Box, 5 Boxes/Case, Sterile, Low-retention, Filtered
SDP
Supplier:  Mascon Medical FSTIPS100
Encompass_Preferred
Mascon Medical™ offers an extensive line of compatible robotic tips for Hamilton, Eppendorf™, Rainin, and Beckman automation systems. These high-precision tips are manufactured to guarantee a perfect fit and extremely accurate dispensing behavior. Available in non-filtered and filtered. Nonpyrogenic. Conductive Polypropylene tips are available in 96-well nested tip rack.Features:•96 Tip/Nested Tip Rack Format•Meets ISO13485 quality management system standard•Nonpyrogenic•Low-retention•UniversalSpecifications:•Volume: 100µL•Qty. / Case: 4800•Size: 50.3mm•Tip Format: 96 Tips/Rack•Material: Polypropylene•Filter: Yes•Sterile: Yes•Packaging: Nested Tip Rack, 10 Racks/Pack, 5 Packs /Carton•Packaging Format: 50 Racks/Carton
Catalog No. NC2458377