Learn More
Biochain Institute Inc Universal Protein Lysate: Human Normal Tissues, 4x0.5 mg/PK
SDP
Supplier:  Biochain Institute Inc P4234565
Encompass_Preferred
BioChain's Universal Protein Lysate is the total protein isolated from a variety of tissue sources including human, monkey, dog, mouse, and rat, to ensure the broadest coverage of proteins. It can be used as controls for comparison of protein expression and for antibody microarray study. Our product is manufactured on an industrial scale to minimize lot-to-lot variation.
Catalog No. 50-184-225