Learn More
Hettich Centrifuges UNIVERSAL320R,(28)13(20)16,Package
SDP
Supplier:  Hettich Centrifuges 320RBLOOD4BC
Encompass_Preferred
Hettich UNIVERSAL 320R BLOOD TUBE PACKAGE 4 BIO-CONTAINMENT, (1) Centrifuge, (1) Horizontal Rotor, (4) Buckets, (4) Adapters 13x 75/ 100 mm, (4) Adapters 16 x 75/ 100 mm, (4) Bio-containment Lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7636
Only null left
Add to Cart
Request a Quote