Learn More
Sigma Aldrich Fine Chemicals Biosciences Unix SEC-200 UHPLC Column, 15 cm x 4.6 mm, 1.8 um particle size
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z777303
Encompass
Utilizing proprietary surface technologies, Unix SEC-200 phase is made of uniform, hydrophilic, and neutral nanometer thick films chemically bonded to high purity and mechanically stabilized silica with the particle size of 1.8 um. The combination of small particle size and large pore volume of Unix SEC-300 renders the highest separation effciency and resolution of analytes. The well controlled surface chemisty results in excellent lot-to-lot reproducibility. The unique bonding chemistry, coupled with the maximized bonding density, allows Unix SEC-200 to provide high stability and negligible non-specific interactions. Typical applications for the Unix SEC-200 columns include separation and analysis of biological molecules and water-soluble polymers.
Catalog No. NC1726099