Learn More
Sterlitech Corporation PTFE Unlaminated Membrane Filters, Hydrophilic, 0.1 Micron, 13mm, 100/Pk
SDP
Supplier:  Sterlitech Corporation H010A013A
Encompass_Preferred
Durable, efficient, and broadly compatible, these pure, hydrophilic PTFE membranes are ideal for use with HPLC seperations, aqueous/organic solvents, and many of your other filtration needs. Pack of 100. Pore size: 0.10 microns. Diameter: 13 mm.Summary of Features:Maximum chemical and pH resistanceHigh flow rates with minimal aqueous extractables (<0.3wt%)Optically clear when wet with waterUnlaminated Ideal for HPLC and other mixtures of aqueous and organic solvents
Catalog No. NC3665966