Learn More
Cayman Chemical Unoproston Isopropyl esTR 10mg
SDP
Supplier:  Cayman Chemical 1668110 MG
Encompass_Preferred
An ocular hypotensive agent, a prodrug form of unoprostone, and a derivative of PGF2α; inhibits endothelin-1-induced contractions in isolated porcine retinal arterioles at 1 nM; inhibits indomethacin-induced increases in intraocular pressure and increases PGE2 levels in the aqueous humor and aqueous humor outflow in rabbits when applied topically at 0.12% v/v
Catalog No. 50-296-5472