Learn More
Enzo Life Sciences AMPIPROBE UP/Mg/Mh Extraction Controls (500µl)
SDP
Supplier:  Enzo Life Sciences ENZGEN2100500
Encompass_Preferred
The AMPIPROBE UP/Mg/Mh Extraction Controls are designed to be used during the genomic DNA extraction of Ureaplasma spp. (Ureaplasma parvum and Ureaplasma urealyticum), Mycoplasma genitalium, and Mycoplasma hominis. All performance specifications must be validated by end-user and will vary based on chosen extraction methods. Eluent from extraction can be used as extraction control for AMPIPROBE UP/Mg/Mh Assay kit (ENZ-GEN209). Plasmids in Extraction Controls are present at higher concentrations compared to assay controls included in the qPCR kit.  Extra care should be taken to avoid contamination. Applications: qPCR. Species reactivity: Mycoplasma hominis, Mycoplasma genitalium, Ureaplasma spp.
Catalog No. 50-201-3768