Learn More
Waters Corp Waters UPC2 Standards Kit, Multi-Component, in 2-Propanol
Supplier:  Waters Corp 176002811
Encompass_Preferred
The UPC2 Standards Kit is a reference kit designed for use with ACQUITY UPC2 Systems. It provides a convenient solution for analytical calibration and system suitability, containing a precisely formulated set of standards in 2-propanol.
  • Contains 1.0 mg/mL caffeine in 2-propanol, supplied in a 2 mL volume.
  • Includes a mixture of 1 mg/mL coumarin, 1 mg/mL flavone, 2 mg/mL caffeine, 1 mg/mL thymine, and 2 mg/mL prednisone in 2-propanol, supplied in a 1 mL volume.
  • Designed for analytical calibration and system suitability.
  • Streamlines purchasing with customized quotes.
Catalog No. 50-465-918