Learn More
ATCC UPCI:SCC090; Squamous Cell Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL3239
Encompass
UPCI:SCC090 is a squamous cell line that was isolated in 1994 from a 46-year-old, white male patient with squamous cell carcinoma, grade 3, pathological stage: T2N0. These cells were isolated from the base of the tongue. They are positive for Human Papilloma Virus (HPV). They have no TP53 mutations as assayed by sequencing the 5-8 exons of TP53. UPCI:SCC090 showed no amplification of chromosomal band 11q13 using FISH. The patient had a recurrence about 1 year later sited at the hypopharynx. It is ATCC No. CRL-3240 UPCI:SCC152. This cell line was deposited by Susanne M. Gollin, University of Pittsburgh, and can be used in studying the initiation, cancerization, prognosis, intervention and treatment of oral cancers.
Catalog No. 50-238-4854