Learn More
Genetex Inc UPK1A Monoclonal Antibody [MSVA-735M] HistoMAX™ (GTX04462) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04462
Encompass_Preferred
The UPK1A antibody [MSVA-735M] HistoMAX™ (GTX04462) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human mucosa from rectum tissue using GTX04462 UPK1A antibody [MSVA-735M] HistoMAX. Complete absence of UPK1A immunostaining in rectal mucosa.
Catalog No. NC3667743