Learn More
Meso Scale Discovery UPLEX Angiogenesis Combo 1 hu
SDP
Supplier:  Meso Scale Discovery K15339K2
Encompass
The U-PLEX Angiogenesis Combo 1 Human Kit by Meso Scale Discovery is a multiplex immunoassay kit for measuring VEGF-A VEGF-C VEGF-D and PlGF biomarkers in human serum plasma and cell culture supernatants It uses MSD s SECTOR technology offering high sensitivity broad dynamic range low sample volume and a 2 5-hour protocol The kit includes a 96-well plate capture and detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer suitable for studying angiogenesis in cancer cardiovascular and inflammatory diseases
Catalog No. 50-000-81518
Add to Cart
Request a Quote