Learn More
Meso Scale Discovery UPLEX Angiogenesis Combo 1 hu
SDP
Supplier:  Meso Scale Discovery K15339K1
Encompass
The U-PLEX Angiogenesis Combo 1 Human Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous measurement of VEGF-A VEGF-C VEGF-D and PlGF in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The electrochemiluminescent assay takes 3 5 hours to complete and has a detection range of 16-10 000 pg/mL for VEGF-A 31-20 000 pg/mL for VEGF-C 16-10 000 pg/mL for VEGF-D and 7 8-5 000 pg/mL for PlGF It allows sensitive and accurate quantification of angiogenic factors
Catalog No. 50-000-82142