Learn More
Meso Scale Discovery UPLEX CART Cell Combo 1 hu
SDP
Supplier:  Meso Scale Discovery K15338K2
Encompass
The U-PLEX CAR-T Cell Combo 1 Human Kit from Meso Scale Discovery is a multiplex immunoassay for detecting human cytokines and chemokines in CAR-T cell therapy research It features a 96-well plate with capture antibodies for 10 analytes including IL-2 IL-4 IL-6 IL-10 IL-17A IFN- TNF- GM-CSF IP-10 and MCP-1 Using electrochemiluminescent detection it offers high sensitivity and a broad dynamic range The kit includes standards detection antibodies buffers and requires an MSD instrument plate reader for quantifying immune markers in various samples
Catalog No. 50-000-78266
Only null left
Add to Cart
Request a Quote