Learn More
Meso Scale Discovery UPLEX CART Cell Combo 1 hu
SDP
Supplier:  Meso Scale Discovery K15338K4
Encompass
The U-PLEX CAR-T Cell Combo 1 Human Kit from Meso Scale Diagnostics provides reagents for measuring cytokines and chemokines in CAR-T cell therapy research It includes assays for IL-2 IL-4 IL-6 IL-8 IL-10 IL-17A IFN- TNF- MCP-1 IP-10 MIG and Eotaxin-3 using electrochemiluminescent detection technology The kit contains plates with capture antibodies calibrators detection antibodies read buffer and wash buffer enabling researchers to profile CAR-T cell responses and understand cytokine release syndrome
Catalog No. 50-000-81885
Add to Cart
Request a Quote