Learn More
Meso Scale Discovery UPLEX Chemokine Combo 2 hu
SDP
Supplier:  Meso Scale Discovery K15046K4
Encompass
The Meso Scale Discovery U-PLEX Chemokine Combo 2 Human Kit is a multiplex immunoassay that simultaneously measures the levels of 10 human chemokines CCL2 CCL3 CCL4 CCL5 CCL11 CXCL1 CXCL8 CXCL9 CXCL10 and CXCL12 This assay utilizes MSD s proprietary electrochemiluminescent detection technology and is designed for use with serum plasma and cell culture supernatant samples The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals This assay allows high-throughput sensitive and reproducible quantification of multiple chemokines from a small sample volume
Catalog No. 50-000-81672
Only null left
Add to Cart
Request a Quote