Learn More
Meso Scale Discovery UPLEX Chemokine Combo 2 ms
SDP
Supplier:  Meso Scale Discovery K15319K4
Encompass
The U-PLEX Chemokine Combo 2 Mouse is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous quantitative measurement of 10 mouse chemokines (CCL2 CCL3 CCL4 CCL5 CCL11 CXCL1 CXCL2 CXCL5 CXCL9 CXCL10) from cell culture supernatants serum plasma (EDTA heparin) and brain tissue lysates The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies and calibrators Sample incubation time is 2 hours
Catalog No. 50-000-80416
Add to Cart
Request a Quote