Learn More
Meso Scale Discovery UPLEX Development Pack3842ASY
SDP
Supplier:  Meso Scale Discovery K25227N1
Encompass
The Meso Scale Discovery U-PLEX Development Pack 384-Well 2-Assay is a multiplex immunoassay kit for developing custom assays on the MSD platform It contains a 384-well MULTI-ARRAY plate 2 vials of SULFO-TAG NHS-Ester labeling reagent blocking solution read buffer 10X wash buffer and plate seals The kit allows for custom coupling of capture antibodies to 2 distinct spots per well Coupled with MSD SULFO-TAG detection antibodies this enables development of 2-plex assays for biomarker quantification The kit provides the essential components for assay development excluding antibodies and analyte standards
Catalog No. 50-000-81041