Learn More
Meso Scale Discovery UPLEX Development Pack7ASY ate
SDP
Supplier:  Meso Scale Discovery K15232N2
Encompass
The Meso Scale Discovery U-PLEX Development Pack 7 Assay is a multiplex immunoassay kit that allows users to develop custom assays for up to 7 biomarkers It contains a 10-plate MULTI-ARRAY 96-well small-spot plate SULFO-TAG NHS-Ester labels Read Buffer Sample Diluent Wash Buffer Blocker A and detection antibodies for 7 analytes This flexible and customizable kit enables detection of protein biomarkers for life science research applications such as cancer immunology neurobiology and cardiovascular disease With electrochemiluminescence technology it offers high sensitivity up to 5 logs of dynamic range and low sample volume requirements
Catalog No. 50-000-78572