Learn More
Meso Scale Discovery UPLEX Development Pack9ASY ate
SDP
Supplier:  Meso Scale Discovery K15234N1
Encompass
The Meso Scale Discovery U-PLEX Development Pack 9 Assay is a multiplex immunoassay kit that allows users to develop custom assays for up to 9 biomarkers It contains a 9-spot MULTI-ARRAY plate reagents and buffers needed to develop and run the assay calibrators and a vial of Read Buffer T The electrochemiluminescent detection method enables high sensitivity and a large dynamic range This product enables researchers to measure biomarkers of interest for their specific applications With optimization limits of detection in the pg/mL range are achievable
Catalog No. 50-000-78451