Learn More
Meso Scale Discovery UPLEX Development Pack9ASY ate
SDP
Supplier:  Meso Scale Discovery K15234N4
Encompass
The Meso Scale Discovery U-PLEX Development Pack 9 Assay is a multiplex immunoassay kit that allows users to develop custom assays for up to 9 biomarkers It contains the components needed to develop and optimize immunoassays on the Meso Scale Discovery electrochemiluminescent detection platform The kit includes assay plates linkers detection antibodies buffers calibrators and reagents for developing and optimizing custom assays It allows flexibility in choosing analyte targets and antibodies while leveraging MSD s sensitive electrochemiluminescent detection system This development kit enables creation of custom multiplex assays for biomarker research
Catalog No. 50-000-82067
Only null left
Add to Cart
Request a Quote