Learn More
Meso Scale Discovery U-PLEX Diabetes Combo 2 (hu) SECTOR (1 PL)
SDP
Supplier:  Meso Scale Discovery K15275K1
Encompass
U-PLEX Combos offer popular panels of biomarkers that have been grouped together for convenient ordering. They have been tested for multiplex compatibility and are fulfilled as a kit. The U-PLEX Diabetes Combo 2 (human) contains assays for seven well-established metabolic analytes that are associated with diabetes (Table 1). The assays can be used for quantitative measurements in the sample types listed in the "Specifications" tab below, and offer detection limits (LLODs) in the sub- to low pg/mL range.
Catalog No. NC3786777