Learn More
Meso Scale Discovery UPLEX HUM ApoA1 ASYQP 25 PL
SDP
Supplier:  Meso Scale Discovery K151S9K24
Encompass
The Meso Scale Discovery U-PLEX Human ApoA1 Assay Kit is a multiplex immunoassay for measuring Apolipoprotein A1 (ApoA1) in human serum plasma and cell culture supernatant It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a 96-well plate antibodies read buffer calibrator and controls Sensitivity is 0 3 ng/mL with a quantification limit of 2500 ng/mL It requires 50 L of sample and takes about 3 5 hours This kit supports research on cardiovascular diseases and lipid metabolism
Catalog No. 50-000-79179
Add to Cart
Request a Quote