Learn More
Meso Scale Discovery UPLEX HUM APRIL/TNFSF13 ASYQP
SDP
Supplier:  Meso Scale Discovery K151AGUK21
Encompass
The Meso Scale Discovery U-PLEX Human APRIL (TNFSF13) Assay is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of APRIL in human serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance It allows for the accurate quantification of human APRIL levels in approximately 4 hours
Catalog No. 50-000-81584