Learn More
Meso Scale Discovery UPLEX HUM BCMA/TNFRSF17 ASYQP
SDP
Supplier:  Meso Scale Discovery K151D8K21
Encompass
The Meso Scale Discovery U-PLEX Human BCMA (TNFRSF17) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of BCMA in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results for BCMA levels This assay can be used to monitor BCMA levels in research studies focused on multiple myeloma autoimmune diseases and other conditions involving BCMA The assay has high sensitivity a broad dynamic range and requires a low sample volume
Catalog No. 50-000-82058