Learn More
Meso Scale Discovery UPLEX HUM BDNF ASY384well
SDP
Supplier:  Meso Scale Discovery K2516WK4
Encompass
The Meso Scale Discovery U-PLEX Human BDNF 384-Well Assay Kit is a multiplex immunoassay that measures brain-derived neurotrophic factor (BDNF) in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This 384-well plate assay allows high throughput analysis of BDNF an important biomarker of neurological function and disease The kit includes a BDNF standard curve and all necessary reagents and plates for quantitative measurement of BDNF levels
Catalog No. 50-000-80999
Only null left
Add to Cart
Request a Quote