Learn More
Meso Scale Discovery UPLEX HUM CD27 ASY384well
SDP
Supplier:  Meso Scale Discovery K251G7K4
Encompass
The Meso Scale Discovery U-PLEX Human CD27 384-Well Assay Kit is a multiplex immunoassay that measures CD27 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay for high throughput analysis of CD27 The assay has high sensitivity wide dynamic range and requires low sample volume It includes a CD27 standard curve and reagents for 384 tests This assay kit can be used for immunology and oncology research applications involving CD27
Catalog No. 50-000-79408
Add to Cart
Request a Quote