Learn More
Meso Scale Discovery UPLEX HUM CD40L soluble ASY384
SDP
Supplier:  Meso Scale Discovery K251L7K4
Encompass
The Meso Scale Discovery U-PLEX Human CD40L (soluble) 384-well assay is a multiplex immunoassay panel that measures soluble CD40 ligand in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This 30-minute assay requires only 25 L of sample and has a lower limit of detection of 45 3 pg/mL It is available as a kit providing a 384-well plate coated with capture antibodies detection antibodies calibrators and controls
Catalog No. 50-000-80467
Only null left
Add to Cart
Request a Quote