Learn More
Meso Scale Discovery UPLEX HUM Comp factor D ASY384
SDP
Supplier:  Meso Scale Discovery K251K9K2
Encompass
The Meso Scale Discovery Human Complement Factor D Singleplex Assay is a quantitative electrochemiluminescent immunoassay designed to measure levels of complement factor D in human serum plasma and other biological fluids This assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The assay has high specificity for human complement factor D with no observed cross-reactivity It is available as a singleplex assay in a 96-well plate format
Catalog No. 50-000-79433
Add to Cart
Request a Quote