Learn More
Meso Scale Discovery UPLEX HUM Comp factor D ASYQP
SDP
Supplier:  Meso Scale Discovery K151K9K21
Encompass
The Meso Scale Discovery U-PLEX Human Complement Factor D Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of Complement Factor D protein in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals The assay requires 50 L of sample and can perform up to 80 tests across two plates
Catalog No. 50-000-80797