Learn More
Meso Scale Discovery UPLEX HUM Complement C9 ASY
SDP
Supplier:  Meso Scale Discovery K151U9K4
Encompass
The Meso Scale Discovery U-PLEX Human Complement C9 Assay Kit quantifies Complement C9 in human serum plasma urine cell culture supernatants and other samples This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection of C9 levels in the pg/mL range The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators controls diluents wash buffer read buffer and plate seals The assay takes about 3 5 hours enabling high-throughput C9 quantification with low sample volume
Catalog No. 50-000-80508
Only null left
Add to Cart
Request a Quote