Learn More
Meso Scale Discovery UPLEX HUM Cystatin C ASY
SDP
Supplier:  Meso Scale Discovery K151M9K4
Encompass
The Meso Scale Discovery U-PLEX Human Cystatin C Assay Kit is an electrochemiluminescent immunoassay for measuring cystatin C in human serum plasma and urine It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection from small samples The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and diluent It measures cystatin C from 0 156 ng/mL to 10 ng/mL offering a simple rapid and robust tool for research and clinical applications
Catalog No. 50-000-79958
Only null left
Add to Cart
Request a Quote