Learn More
Meso Scale Discovery UPLEX HUM Cystatin C ASY384
SDP
Supplier:  Meso Scale Discovery K251M9K4
Encompass
The Meso Scale Discovery U-PLEX Human Cystatin C 384-Well Assay is an electrochemiluminescent immunoassay designed for the quantitative measurement of Cystatin C in human serum plasma and other biological fluids This multiplex assay kit provides a 384-well plate pre-coated with capture antibodies for Cystatin C along with detection antibodies and calibrators The assay has a typical sensitivity of 0 196 ng/mL and an assay range from 0 049 to 25 ng/mL It uses MSD s proprietary SULFO-TAG labels that emit light when electrochemically stimulated to enable high sensitivity This allows for the rapid and accurate quantification of Cystatin C from small sample volumes
Catalog No. 50-000-80690
Only null left
Add to Cart
Request a Quote