Learn More
Meso Scale Discovery UPLEX HUM Eotaxin ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K151UDK4
Encompass
The Meso Scale Discovery U-PLEX Human Eotaxin Assay Kit quantitatively measures human Eotaxin (CCL11) in various biological samples This electrochemiluminescent multiplex immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection down to 0 04 pg/mL with minimal sample volume The assay offers high specificity with no cross-reactivity and includes reagents for a 96-well plate enabling high-throughput analysis This advanced kit allows precise quantification of human Eotaxin across diverse sample types
Catalog No. 50-000-80407
Add to Cart
Request a Quote