Learn More
Meso Scale Discovery UPLEX HUM EPO ASY384well
SDP
Supplier:  Meso Scale Discovery K251VXK4
Encompass
The Meso Scale Discovery U-PLEX Human EPO 384-Well Assay is an electrochemiluminescent immunoassay for the quantitative measurement of erythropoietin (EPO) in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results in a 384-well format The assay has a broad dynamic range of up to 10 000 pg/mL and requires as little as 25 L of sample Combined with the efficiency of 384-well plates it enables high-throughput screening and monitoring of EPO levels
Catalog No. 50-000-80664
Only null left
Add to Cart
Request a Quote