Learn More
Meso Scale Discovery UPLEX HUM ESelectin ASY384well
SDP
Supplier:  Meso Scale Discovery K251AGVK2
Encompass
The Meso Scale Discovery U-PLEX Human E-Selectin Assay is a 384-well multiplex immunoassay kit for measuring E-Selectin in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results The assay requires a small sample volume and takes about 3 5 hours It includes a 384-well plate with capture antibodies detection antibodies calibrators and controls The kit performs up to 380 tests and lasts 9 months at 2-8 C enabling efficient E-Selectin quantification
Catalog No. 50-000-80817
Add to Cart
Request a Quote