Learn More
Meso Scale Discovery UPLEX HUM Factor VII ASY384
SDP
Supplier:  Meso Scale Discovery K251N9K2
Encompass
The Meso Scale Discovery U-PLEX Human Factor VII 384-Well Assay is an electrochemiluminescent immunoassay designed for the quantitative measurement of Factor VII in human serum plasma and cell culture supernatants This multiplex assay allows high-throughput screening and profiling of Factor VII levels with high sensitivity and broad dynamic range The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and controls for generating standard curves and assessing assay performance This assay provides a rapid reproducible method for measuring Factor VII levels in research and drug discovery applications
Catalog No. 50-000-80444
Add to Cart
Request a Quote