Learn More
Meso Scale Discovery UPLEX HUM Factor VII ASYQP
SDP
Supplier:  Meso Scale Discovery K151N9K24
Encompass
The Meso Scale Discovery U-PLEX Human Factor VII Assay Kit is a multiplex immunoassay for measuring Factor VII in human serum plasma and cell culture supernatants It uses electrochemiluminescent detection for high sensitivity and a broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent tags calibrators and diluent Sensitivity is 0 03 ng/mL with an upper quantification limit of 1500 ng/mL Compatible with Meso Scale Discovery SECTOR Imagers and QuickPlex SQ 120 it ensures reliable Factor VII quantification
Catalog No. 50-000-82266
Add to Cart
Request a Quote