Learn More
Meso Scale Discovery UPLEX HUM FGF23 ASYQP 25 PL
SDP
Supplier:  Meso Scale Discovery K1516EK24
Encompass
The U-PLEX Human FGF-23 Assay (K1516EK) is a multiplex electrochemiluminescence immunoassay designed to quantitatively measure fibroblast growth factor 23 (FGF-23) levels in human serum plasma and cell culture samples This research-grade assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity detection with a wide dynamic range The kit includes pre-coated plates detection antibodies and calibration standards for reliable reproducible results in clinical research and biomarker studies
Catalog No. 50-000-80312
Only null left
Add to Cart
Request a Quote