Learn More
Meso Scale Discovery UPLEX HUM FLT3L ASY384well
SDP
Supplier:  Meso Scale Discovery K251XFK4
Encompass
The Meso Scale Discovery U-PLEX Human FLT3L 384-Well Assay Kit is a multiplex immunoassay that measures FLT3L in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls It allows simultaneous measurement of FLT3L and other biomarkers The assay has a typical lower limit of detection of 0 06 pg/mL and an upper limit of quantification of 2500 pg/mL for FLT3L
Catalog No. 50-000-80705
Only null left
Add to Cart
Request a Quote