Learn More
Meso Scale Discovery UPLEX HUM Fractalkine ASY384
SDP
Supplier:  Meso Scale Discovery K251VCK4
Encompass
The Meso Scale Discovery U-PLEX Human Fractalkine 384-Well Assay Kit is a multiplex immunoassay that measures human Fractalkine in cell culture supernatants serum plasma and other biological fluids This kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay for high throughput screening The assay has high sensitivity and wide dynamic range This product requires a Meso Scale Discovery instrument such as the MESO QuickPlex SQ 120 for detection Each kit provides reagents for one 384-well plate and can test 40 samples in duplicate
Catalog No. 50-000-79475
Only null left
Add to Cart
Request a Quote